missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-3-(Boc-amino)pyrrolidine, 97%

Catalog No. AAH5241403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5241403 1 g
AAH5241406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5241403 Supplier Thermo Scientific Chemicals Supplier No. H5241403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 99724-19-3 | C9H18N2O2 | 186.255 g/mol

(±)-3-(Boc-amino)pyrrolidine is the homochiral starting materials utilised in the synthesis of all of the chiral compounds. Reactant in the peptide activated G-protein coupled receptors test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-3-(Boc-amino)pyrrolidine is the homochiral starting materials utilised in the synthesis of all of the chiral compounds. Reactant in the peptide activated G-protein coupled receptors test.

Solubility
Soluble in Methanol(almost transparency).

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Chemical Identifiers

CAS 99724-19-3
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.255
MDL Number MFCD00059040
InChI Key DQQJBEAXSOOCPG-UHFFFAOYSA-N
Synonym 3-boc-aminopyrrolidine, tert-butyl pyrrolidin-3-ylcarbamate, 3-tert-butoxycarbonylamino pyrrolidine, 3-n-boc-aminopyrrolidine, 3-n-boc-amino pyrrolidine, 3-boc-amino pyrrolidine, tert-butyl n-pyrrolidin-3-yl carbamate, 3-tert-butoxycarbonylamino-pyrrolidine, 3-aminopyrrolidine, 3-boc protected, 3-tert-butoxycarbonyl amino pyrrolidine
PubChem CID 2757234
IUPAC Name tert-butyl N-pyrrolidin-3-ylcarbamate
SMILES CC(C)(C)OC(=O)NC1CCNC1

Specifications

Melting Point 54°C
Solubility Information Soluble in Methanol(almost transparency).
Quantity 1 g
Formula Weight 186.25
Sensitivity Air Sensitive
Percent Purity 97%
Chemical Name or Material (+/-)-3-(Boc-amino)pyrrolidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only