Learn More
3-(Boc-amino)azetidine, 98%
CAS: 91188-13-5 | C8H16N2O2 | 172.23 g/mol
$154.99 - $270.10
Chemical Identifiers
| CAS | 91188-13-5 |
|---|---|
| Molecular Formula | C8H16N2O2 |
| Molecular Weight (g/mol) | 172.23 |
| MDL Number | MFCD01861759 |
| InChI Key | NEMXVXVJGXZDRR-UHFFFAOYSA-N |
| Synonym | tert-butyl azetidin-3-ylcarbamate, 3-n-boc-amino-azetidine, 3-boc-aminoazetidine, 3-n-boc-amino azetidine, tert-butyl n-azetidin-3-yl carbamate, 3-n-boc-aminoazetidine, 3-boc-amino azetidine, azetidin-3-yl-carbamic acid tert-butyl ester, 3-boc-amino-azetidine, 3-tert-butoxycarbonylamino azetidine |
| PubChem CID | 1519405 |
| IUPAC Name | tert-butyl N-(azetidin-3-yl)carbamate |
| SMILES | CC(C)(C)OC(=O)NC1CNC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH59556MD
|
Thermo Scientific Chemicals
H59556MD |
250 mg |
Each for $154.99
|
|
|||||
|
AAH59556ME
|
Thermo Scientific Chemicals
H59556ME |
500 mg |
Each for $270.10
|
|
|||||
Description
It is an intermediate in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an intermediate in organic syntheses.
Solubility
Soluble in water. (5g/100ml)
Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
Chemical Identifiers
| 91188-13-5 | |
| 172.23 | |
| NEMXVXVJGXZDRR-UHFFFAOYSA-N | |
| 1519405 | |
| CC(C)(C)OC(=O)NC1CNC1 |
| C8H16N2O2 | |
| MFCD01861759 | |
| tert-butyl azetidin-3-ylcarbamate, 3-n-boc-amino-azetidine, 3-boc-aminoazetidine, 3-n-boc-amino azetidine, tert-butyl n-azetidin-3-yl carbamate, 3-n-boc-aminoazetidine, 3-boc-amino azetidine, azetidin-3-yl-carbamic acid tert-butyl ester, 3-boc-amino-azetidine, 3-tert-butoxycarbonylamino azetidine | |
| tert-butyl N-(azetidin-3-yl)carbamate |
Specifications
| 91188-13-5 | |
| C8H16N2O2 | |
| tert-butyl azetidin-3-ylcarbamate, 3-n-boc-amino-azetidine, 3-boc-aminoazetidine, 3-n-boc-amino azetidine, tert-butyl n-azetidin-3-yl carbamate, 3-n-boc-aminoazetidine, 3-boc-amino azetidine, azetidin-3-yl-carbamic acid tert-butyl ester, 3-boc-amino-azetidine, 3-tert-butoxycarbonylamino azetidine | |
| NEMXVXVJGXZDRR-UHFFFAOYSA-N | |
| 1.06 | |
| 250 mg | |
| 116°C (241°F) | |
| 172.23 | |
| 3-(Boc-amino)azetidine |
| 268°C | |
| MFCD01861759 | |
| Soluble in water. (5g/100ml) | |
| CC(C)(C)OC(=O)NC1CNC1 | |
| tert-butyl N-(azetidin-3-yl)carbamate | |
| 172.23 | |
| 1519405 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only