missing translation for 'onlineSavingsMsg'
Learn More

3-(BOC-amino)-1-propanol, 97%

Catalog No. AC399800100
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC399800100 10 mL
AC399800500 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399800100 Supplier Thermo Scientific Chemicals Supplier No. 399800100
Only null left

CAS: 58885-58-8 | C8H17NO3 | 175.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58885-58-8
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.23
MDL Number MFCD00191883
InChI Key XDJCYKMWJCYQJM-UHFFFAOYSA-N
Synonym 3-boc-amino-1-propanol, tert-butyl n-3-hydroxypropyl carbamate, boc-beta-ala-ol, 3-tert-butoxycarbonylamino-1-propanol, tert-butyl 3-hydroxypropylcarbamate, 3-n-boc-amino propanol, tert-butyl 3-hydroxypropyl carbamate, n-3-hydroxypropyl carbamic acid tert-butyl ester, 3-hydroxy-propyl-carbamic acid tert-butyl ester
PubChem CID 3838134
IUPAC Name tert-butyl N-(3-hydroxypropyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCO

Specifications

Density 1.0250g/mL
Flash Point 109°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula HO(CH2)3NHCO2C(CH3)3
Quantity 10 mL
Specific Gravity 1.025
Formula Weight 175.23
Percent Purity 97%
Chemical Name or Material 3-(BOC-amino)-1-propanol
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only