missing translation for 'onlineSavingsMsg'
Learn More

3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulfonic acid, 99%

Catalog No. p-7037448
Change view
Click to view available options
Quantity:
100 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5618722 100 g
AAH5618730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5618722 Supplier Thermo Scientific Chemicals Supplier No. H5618722

CAS: 68399-80-4 | C7H17NO6S | 243.274 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68399-80-4
Molecular Formula C7H17NO6S
Molecular Weight (g/mol) 243.274
MDL Number MFCD00038353
InChI Key XCBLFURAFHFFJF-UHFFFAOYSA-N
Synonym dipso, 3-bis 2-hydroxyethyl amino-2-hydroxypropane-1-sulfonic acid, 3-n,n-bis 2-hydroxyethyl amino-2-hydroxypropanesulfonic acid, 3-n,n-bis 2-hydroxyethyl amino-2-hydroxy-1-propanesulfonic acid, 1-propanesulfonic acid, 3-bis 2-hydroxyethyl amino-2-hydroxy, 3-bis 2-hydroxyethyl amino-2-hydroxypropanesulfonic acid, 3-bis 2-hydroxyethyl amino-2-hydroxy-1-propanesulfonic acid, n,n-bis 2-hydroxyethyl-3-amino-2-hydroxypropanesulfonic acid
PubChem CID 100210
IUPAC Name 3-[bis(2-hydroxyethyl)amino]-2-hydroxypropane-1-sulfonic acid
SMILES C(CO)N(CCO)CC(CS(=O)(=O)O)O

Specifications

Quantity 100 g
Beilstein 2105454
Formula Weight 261.29
Percent Purity 99%
Chemical Name or Material 3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulfonic acid
EINECSNumber 269-992-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only