missing translation for 'onlineSavingsMsg'
Learn More

3-Biphenylcarboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 716-76-7
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00045846
InChI Key XNLWJFYYOIRPIO-UHFFFAOYSA-N
Synonym biphenyl-3-carboxylic acid, 3-biphenylcarboxylic acid, 1,1'-biphenyl-3-carboxylic acid, diphenyl-3-carboxylic acid, wln: qvr cr, 3-biphenylcarboxylicacid, m-phenylbenzoic acid, 3-phenyl benzoic acid
PubChem CID 12854
IUPAC Name 3-phenylbenzoic acid
SMILES C1=CC=C(C=C1)C2=CC(=CC=C2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC371770010
View Documents Promotion Details
N/A 1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC371770050
View Documents Promotion Details
N/A 5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

716-76-7
198.22
XNLWJFYYOIRPIO-UHFFFAOYSA-N
12854
C1=CC=C(C=C1)C2=CC(=CC=C2)C(=O)O
C13H10O2
MFCD00045846
biphenyl-3-carboxylic acid, 3-biphenylcarboxylic acid, 1,1'-biphenyl-3-carboxylic acid, diphenyl-3-carboxylic acid, wln: qvr cr, 3-biphenylcarboxylicacid, m-phenylbenzoic acid, 3-phenyl benzoic acid
3-phenylbenzoic acid

Specifications

716-76-7
100.0
97%
C6H5C6H4COOH
09, IV, 2478
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,acetone,diethyl ether,hexane,soluble in benzene,petroleum ether,alkali
C1=CC=C(C=C1)C2=CC(=CC=C2)C(=O)O
198.22
198.22
3-Biphenylcarboxylic acid
96.0
Authentic
C13H10O2
MFCD00045846
biphenyl-3-carboxylic acid, 3-biphenylcarboxylic acid, 1,1'-biphenyl-3-carboxylic acid, diphenyl-3-carboxylic acid, wln: qvr cr, 3-biphenylcarboxylicacid, m-phenylbenzoic acid, 3-phenyl benzoic acid
XNLWJFYYOIRPIO-UHFFFAOYSA-N
3-phenylbenzoic acid
12854
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only