missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7193927
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B30565G Supplier TCI America Supplier No. B30565G

Chemical Identifiers

CAS 156682-54-1
Molecular Formula C13H13BO3
Molecular Weight (g/mol) 228.05
MDL Number MFCD02093063
InChI Key WIJNYNBSPQMJGO-UHFFFAOYSA-N
PubChem CID 2734313
IUPAC Name [3-(benzyloxy)phenyl]boronic acid
SMILES OB(O)C1=CC(OCC2=CC=CC=C2)=CC=C1

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 5 g
Synonym 3-benzyloxyphenylboronic acid, 3-benzyloxybenzeneboronic acid, 3-benzyloxy phenyl boronic acid, 3-benzyloxy phenylboronic acid, m-benzyloxyphenylboronic acid, 3-benzyloxyphenyl boronic acid, 3-boronophenoxy methyl benzene, 3-benzyl-oxy phenyl boranediol, boronic acid, 3-phenylmethoxy phenyl
Formula Weight 228.05
Physical Form Crystalline Powder
Chemical Name or Material 3-Benzyloxyphenylboronic Acid (contains varying amounts of Anhydride)