Learn More
3-Benzyloxybromobenzene, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 437790100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 3-Benzyloxybromobenzene | |
| 95.0 | |
| Authentic | |
| Glass bottle | |
| 10g | |
| HVWZMGZBJCJDOX-UHFFFAOYSA-N | |
| 1-bromo-3-phenylmethoxybenzene | |
| 2756638 | |
| 96% |
| 53087-13-1 | |
| 100.0 | |
| 95% min. (GC) | |
| C13H11BrO | |
| 1-benzyloxy-3-bromobenzene, 3-benzyloxybromobenzene, benzyl 3-bromophenyl ether, 1-bromo-3-benzyloxybenzene, 3-benzyloxy bromobenzene, 3-bromophenyl benzyl ether, benzene, 1-bromo-3-phenylmethoxy, 1-benzyloxy-3-bromo-benzene, pubchem3065 | |
| C1=CC=C(C=C1)COC2=CC(=CC=C2)Br | |
| 263.13 | |
| 263.13 |
Chemical Identifiers
| 53087-13-1 | |
| 263.13 | |
| 1-benzyloxy-3-bromobenzene, 3-benzyloxybromobenzene, benzyl 3-bromophenyl ether, 1-bromo-3-benzyloxybenzene, 3-benzyloxy bromobenzene, 3-bromophenyl benzyl ether, benzene, 1-bromo-3-phenylmethoxy, 1-benzyloxy-3-bromo-benzene, pubchem3065 | |
| 1-bromo-3-phenylmethoxybenzene |
| C13H11BrO | |
| HVWZMGZBJCJDOX-UHFFFAOYSA-N | |
| 2756638 | |
| C1=CC=C(C=C1)COC2=CC(=CC=C2)Br |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Danger
RUO â Research Use Only