missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3'-Benzyloxyacetophenone, 97%
CAS: 34068-01-4 | C15H14O2 | 226.275 g/mol
$84.56 - $843.82
Chemical Identifiers
| CAS | 34068-01-4 |
|---|---|
| Molecular Formula | C15H14O2 |
| Molecular Weight (g/mol) | 226.275 |
| MDL Number | MFCD00026221 |
| InChI Key | FGQMEAWGAUALJQ-UHFFFAOYSA-N |
| Synonym | 1-3-benzyloxy phenyl ethanone, 3-benzyloxyacetophenone, 3'-benzyloxyacetophenone, 1-3-benzyloxy phenyl ethan-1-one, 1-3-phenylmethoxy phenyl ethan-1-one, 3'-benzyloxy acetophenone, ethanone, 1-3-phenylmethoxy phenyl, 1-acetyl-3-phenylmethoxy benzene |
| PubChem CID | 98689 |
| IUPAC Name | 1-(3-phenylmethoxyphenyl)ethanone |
| SMILES | CC(=O)C1=CC(=CC=C1)OCC2=CC=CC=C2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2508406
|
Thermo Scientific Chemicals
B2508406 |
5 g |
Each for $84.56
|
|
|||||
|
AAB2508414
|
Thermo Scientific Chemicals
B2508414 |
25 g |
Each for $284.22
|
|
|||||
|
AAB2508422
|
Thermo Scientific Chemicals
B2508422 |
100 g |
Each for $843.82
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 34068-01-4 | |
| 226.275 | |
| FGQMEAWGAUALJQ-UHFFFAOYSA-N | |
| 98689 | |
| CC(=O)C1=CC(=CC=C1)OCC2=CC=CC=C2 |
| C15H14O2 | |
| MFCD00026221 | |
| 1-3-benzyloxy phenyl ethanone, 3-benzyloxyacetophenone, 3'-benzyloxyacetophenone, 1-3-benzyloxy phenyl ethan-1-one, 1-3-phenylmethoxy phenyl ethan-1-one, 3'-benzyloxy acetophenone, ethanone, 1-3-phenylmethoxy phenyl, 1-acetyl-3-phenylmethoxy benzene | |
| 1-(3-phenylmethoxyphenyl)ethanone |
Specifications
| 34068-01-4 | |
| MFCD00026221 | |
| 1-3-benzyloxy phenyl ethanone, 3-benzyloxyacetophenone, 3'-benzyloxyacetophenone, 1-3-benzyloxy phenyl ethan-1-one, 1-3-phenylmethoxy phenyl ethan-1-one, 3'-benzyloxy acetophenone, ethanone, 1-3-phenylmethoxy phenyl, 1-acetyl-3-phenylmethoxy benzene | |
| CC(=O)C1=CC(=CC=C1)OCC2=CC=CC=C2 | |
| 226.275 | |
| 226.28 | |
| 3'-Benzyloxyacetophenone |
| C15H14O2 | |
| 5 g | |
| FGQMEAWGAUALJQ-UHFFFAOYSA-N | |
| 1-(3-phenylmethoxyphenyl)ethanone | |
| 98689 | |
| 97% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 251-816-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only