missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxy-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

$250.67 - $787.33

Chemical Identifiers

CAS 874152-84-8
Molecular Formula C19H22BFO3
Molecular Weight (g/mol) 328.19
MDL Number MFCD22419264
InChI Key QLNMPTKMOCZFKS-UHFFFAOYSA-N
Synonym 3-benzyloxy-2-fluorobenzeneboronic acid pinacol ester, 2-3-benzyloxy-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3-benzyloxy-2-fluoro-phenyl-4,4,5,5-tetramethyl 1,3,2 dioxaborolane
PubChem CID 59344877
IUPAC Name 2-(2-fluoro-3-phenylmethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)OCC3=CC=CC=C3)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59049MD
View Documents
Thermo Scientific Chemicals
H59049MD
250 mg
Each for $250.67
Only null left
 
AAH5904903
View Documents
Thermo Scientific Chemicals
H5904903
1 g
Each for $787.33
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

874152-84-8
328.19
QLNMPTKMOCZFKS-UHFFFAOYSA-N
59344877
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)OCC3=CC=CC=C3)F
C19H22BFO3
MFCD22419264
3-benzyloxy-2-fluorobenzeneboronic acid pinacol ester, 2-3-benzyloxy-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3-benzyloxy-2-fluoro-phenyl-4,4,5,5-tetramethyl 1,3,2 dioxaborolane
2-(2-fluoro-3-phenylmethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Specifications

874152-84-8
MFCD22419264
3-benzyloxy-2-fluorobenzeneboronic acid pinacol ester, 2-3-benzyloxy-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3-benzyloxy-2-fluoro-phenyl-4,4,5,5-tetramethyl 1,3,2 dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)OCC3=CC=CC=C3)F
328.19
328.19
3-Benzyloxy-2-fluorobenzeneboronic acid pinacol ester
C19H22BFO3
250 mg
QLNMPTKMOCZFKS-UHFFFAOYSA-N
2-(2-fluoro-3-phenylmethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
59344877
96%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'