missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxy-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog No. AAH59049MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59049MD 250 mg
AAH5904903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59049MD Supplier Thermo Scientific Chemicals Supplier No. H59049MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 874152-84-8
Molecular Formula C19H22BFO3
Molecular Weight (g/mol) 328.19
MDL Number MFCD22419264
InChI Key QLNMPTKMOCZFKS-UHFFFAOYSA-N
Synonym 3-benzyloxy-2-fluorobenzeneboronic acid pinacol ester, 2-3-benzyloxy-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3-benzyloxy-2-fluoro-phenyl-4,4,5,5-tetramethyl 1,3,2 dioxaborolane
PubChem CID 59344877
IUPAC Name 2-(2-fluoro-3-phenylmethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)OCC3=CC=CC=C3)F

Specifications

Quantity 250 mg
Formula Weight 328.19
Percent Purity 96%
Chemical Name or Material 3-Benzyloxy-2-fluorobenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures