missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxazolidine 98.0+%, TCI America™

Catalog No. B13395G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13395G 5 g
B13391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13395G Supplier TCI America Supplier No. B13395G
Only null left

Chemical Identifiers

CAS 13657-16-4
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00059028
InChI Key FQNODHUYZYLTPN-UHFFFAOYSA-N
PubChem CID 319258
IUPAC Name 3-benzyl-1,3-oxazolidine
SMILES C1COCN1CC2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 127°C
Quantity 5 g
Formula Weight 163.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Benzyloxazolidine