missing translation for 'onlineSavingsMsg'
Learn More

3-Benzylidene-2,4-pentanedione, 97%

Catalog No. AAL0062306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0062306 5 g
AAL0062314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0062306 Supplier Thermo Scientific Chemicals Supplier No. L0062306

CAS: 4335-90-4 | C12H12O2 | 188.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4335-90-4
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.23
MDL Number MFCD00014990
InChI Key NYRGMNMVISROGJ-UHFFFAOYSA-N
Synonym 3-benzylidene-2,4-pentanedione, benzalacetylacetone, benzylidene acetylacetone, 3-benzylideneacetylacetone, 2,4-pentanedione, 3-phenylmethylene, 1,1-diacetyl-2-phenylethylene, 2,2-diacetylvinyl benzene, 2-acetyl-1-phenyl but-1-en-3-one, 2,4-pentanedione, 3-benzylidene, 3-benzylidene acetylacetone
PubChem CID 95782
IUPAC Name 3-benzylidenepentane-2,4-dione
SMILES CC(=O)C(=CC1=CC=CC=C1)C(C)=O

Specifications

Density 1.093
Boiling Point 185°C to 188°C (15 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 972849
Formula Weight 188.23
Percent Purity 97%
Chemical Name or Material 3-Benzylidene-2,4-pentanedione
EINECSNumber 224-382-5
RTECSNumber SA1930000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only