missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium Chloride 98.0+%, TCI America™

Catalog No. B122425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B122425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B122425G Supplier TCI America Supplier No. B122425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4568-71-2
Molecular Formula C13H16ClNOS
Molecular Weight (g/mol) 269.79
MDL Number MFCD00011959
InChI Key IWSVLBKHBJGMAA-UHFFFAOYSA-M
Synonym 3-benzyl-5-2-hydroxyethyl-4-methylthiazolium chloride, 3-benzyl-5-2-hydroxyethyl-4-methylthiazol-3-ium chloride, 3-benzyl-5-2-hydroxyethyl-4-methyl-1,3-thiazol-3-ium chloride, thiazolium, 5-2-hydroxyethyl-4-methyl-3-phenylmethyl-, chloride, 2-4-methyl-3-benzyl-1,3-thiazol-5-yl ethan-1-ol, chloride, acmc-1aqyn, 3-benzyl-5-2-hydroxyethyl thazolium chloride, 3-benzyl-4-methyl-5-hydroxyethylthiazolium chloride, 3-benzyl-5-hydroxyethyl-4-methylthiazolium chloride, 3-benzyl-5 2-hydroxyethyl-4-methylthiazolium chloride
PubChem CID 2833352
IUPAC Name 3-benzyl-5-(2-hydroxyethyl)-4-methyl-1,3-thiazol-3-ium chloride
SMILES [Cl-].CC1=C(CCO)SC=[N+]1CC1=CC=CC=C1

Specifications

Melting Point 143°C
Color White-Yellow
Quantity 25 g
Formula Weight 269.79
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium Chloride