missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride, 98%

Catalog No. AC440520050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC440520050 5 g Glass bottle
AC440520250 25 g Glass bottle
AC440521000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC440520050 Supplier Thermo Scientific Chemicals Supplier No. 440520050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4568-71-2 | C13H16ClNOS | 269.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4568-71-2
Molecular Formula C13H16ClNOS
Molecular Weight (g/mol) 269.79
MDL Number MFCD00011959
InChI Key IWSVLBKHBJGMAA-UHFFFAOYSA-M
Synonym 3-benzyl-5-2-hydroxyethyl-4-methylthiazolium chloride, 3-benzyl-5-2-hydroxyethyl-4-methylthiazol-3-ium chloride, 3-benzyl-5-2-hydroxyethyl-4-methyl-1,3-thiazol-3-ium chloride, thiazolium, 5-2-hydroxyethyl-4-methyl-3-phenylmethyl-, chloride, 2-4-methyl-3-benzyl-1,3-thiazol-5-yl ethan-1-ol, chloride, acmc-1aqyn, 3-benzyl-5-2-hydroxyethyl thazolium chloride, 3-benzyl-4-methyl-5-hydroxyethylthiazolium chloride, 3-benzyl-5-hydroxyethyl-4-methylthiazolium chloride, 3-benzyl-5 2-hydroxyethyl-4-methylthiazolium chloride
PubChem CID 2833352
SMILES [Cl-].CC1=C(CCO)SC=[N+]1CC1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 142.0°C to 146.0°C
Color White
Elemental Analysis 56.52 to 59.32% C, 5.06 to 5.32% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: in water:(20°C) soluble.,in ethanol:(20°C) soluble
IUPAC Name 2-(3-benzyl-4-methyl-1,3-thiazol-3-ium-5-yl)ethanol;chloride
Formula Weight 269.8
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-947-6

RUO – Research Use Only