missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride, 98%

Catalog No. AAL0875022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0875022 100 g
AAL0875014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0875022 Supplier Thermo Scientific Chemicals Supplier No. L0875022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4568-71-2 | C13H16ClNOS | 269.79 g/mol

3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride, aliphatic aldehydes undergo the acyloin condensation in the presence of this catalyst and mild base. It acts as a catalyst for the addition of aliphatic and heterocyclic aldehydes to α,β-unsaturated ketones, nitriles and esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride, aliphatic aldehydes undergo the acyloin condensation in the presence of this catalyst and mild base. It acts as a catalyst for the addition of aliphatic and heterocyclic aldehydes to α,β-unsaturated ketones, nitriles and esters.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from water and oxidizing agents. Protect from humidity and water. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 4568-71-2
Molecular Formula C13H16ClNOS
Molecular Weight (g/mol) 269.79
MDL Number MFCD00011959
InChI Key IWSVLBKHBJGMAA-UHFFFAOYSA-M
Synonym 3-benzyl-5-2-hydroxyethyl-4-methylthiazolium chloride, 3-benzyl-5-2-hydroxyethyl-4-methylthiazol-3-ium chloride, 3-benzyl-5-2-hydroxyethyl-4-methyl-1,3-thiazol-3-ium chloride, thiazolium, 5-2-hydroxyethyl-4-methyl-3-phenylmethyl-, chloride, 2-4-methyl-3-benzyl-1,3-thiazol-5-yl ethan-1-ol, chloride, acmc-1aqyn, 3-benzyl-5-2-hydroxyethyl thazolium chloride, 3-benzyl-4-methyl-5-hydroxyethylthiazolium chloride, 3-benzyl-5-hydroxyethyl-4-methylthiazolium chloride, 3-benzyl-5 2-hydroxyethyl-4-methylthiazolium chloride
PubChem CID 2833352
IUPAC Name 2-(3-benzyl-4-methyl-1,3-thiazol-3-ium-5-yl)ethanol;chloride
SMILES [Cl-].CC1=C(CCO)SC=[N+]1CC1=CC=CC=C1

Specifications

Melting Point 142°C to 145°C
Quantity 100 g
Beilstein 3788921
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 269.8
Percent Purity 98%
Chemical Name or Material 3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-947-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only