missing translation for 'onlineSavingsMsg'
Learn More

3-Benzoylthiazolidine-2-thione 98.0+%, TCI America™

Catalog No. B35711G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35711G 1 g
B35715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35711G Supplier TCI America Supplier No. B35711G
Only null left

Chemical Identifiers

CAS 70326-37-3
Molecular Formula C10H9NOS2
Molecular Weight (g/mol) 223.308
InChI Key NWKQMWLTQNBOHI-UHFFFAOYSA-N
PubChem CID 839954
IUPAC Name phenyl-(2-sulfanylidene-1,3-thiazolidin-3-yl)methanone
SMILES C1CSC(=S)N1C(=O)C2=CC=CC=C2

Specifications

Melting Point 109°C
Color Yellow
Quantity 1 g
Formula Weight 223.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Benzoylthiazolidine-2-thione