missing translation for 'onlineSavingsMsg'
Learn More

3-Benzoylpyridine 99.0+%, TCI America™

Catalog No. p-7118405
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B03055G 5 g
B030525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B03055G Supplier TCI America Supplier No. B03055G
Only null left

Chemical Identifiers

CAS 5424-19-1
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD00006394
InChI Key RYMBAPVTUHZCNF-UHFFFAOYSA-N
Synonym 3-benzoylpyridine, phenyl pyridin-3-yl methanone, phenyl 3-pyridyl ketone, methanone, phenyl-3-pyridinyl, pyridine, 3-benzoyl, 3-pyridyl phenyl ketone, ketone, phenyl 3-pyridyl, 3-benzoyl pyridine, phenyl-b-pyridyl ketone, phenyl 3-pyridinyl methanone
PubChem CID 21540
IUPAC Name phenyl(pyridin-3-yl)methanone
SMILES C1=CC=C(C=C1)C(=O)C2=CN=CC=C2

Specifications

Melting Point 39°C
Color White-Yellow
Boiling Point 307°C
Quantity 5 g
Formula Weight 183.21
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Benzoylpyridine
RTECSNumber OB6401000
TSCA No