missing translation for 'onlineSavingsMsg'
Learn More

3-Benzoylpyridine, 98+%

Catalog No. AAA1669522
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1669522 100 g
AAA1669506 5 g
AAA1669514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1669522 Supplier Thermo Scientific Chemicals Supplier No. A1669522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5424-19-1 | C12H9NO | 183.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5424-19-1
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD00006394
InChI Key RYMBAPVTUHZCNF-UHFFFAOYSA-N
Synonym 3-benzoylpyridine, phenyl pyridin-3-yl methanone, phenyl 3-pyridyl ketone, methanone, phenyl-3-pyridinyl, pyridine, 3-benzoyl, 3-pyridyl phenyl ketone, ketone, phenyl 3-pyridyl, 3-benzoyl pyridine, phenyl-b-pyridyl ketone, phenyl 3-pyridinyl methanone
PubChem CID 21540
IUPAC Name phenyl(pyridin-3-yl)methanone
SMILES C1=CC=C(C=C1)C(=O)C2=CN=CC=C2

Specifications

Melting Point 36°C to 40°C
Boiling Point 307°C
Flash Point 150°C (302°F)
Quantity 100 g
Beilstein 120234
Formula Weight 183.21
Percent Purity ≥98%
Chemical Name or Material 3-Benzoylpyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-561-3
RTECSNumber OB6401000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only