missing translation for 'onlineSavingsMsg'
Learn More

3-Benzoylpropionic acid, 98+%

Catalog No. p-7023258
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1663614 25 g
AAA1663622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1663614 Supplier Thermo Scientific Chemicals Supplier No. A1663614
Only null left

CAS: 2051-95-8 | C10H10O3 | 178.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2051-95-8
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00002792
InChI Key KMQLIDDEQAJAGJ-UHFFFAOYSA-N
Synonym 3-benzoylpropionic acid, 4-oxo-4-phenylbutyric acid, 3-benzoylpropanoic acid, beta-benzoylpropionic acid, benzoylpropionic acid, propanoic acid, 3-benzoyl, benzenebutanoic acid, .gamma.-oxo, 3-benzoylpropionicacid, propionic acid, 3-benzoyl, 4-phenyl-4-oxobutyric acid
PubChem CID 72871
ChEBI CHEBI:64437
IUPAC Name 4-oxo-4-phenylbutanoic acid
SMILES OC(=O)CCC(=O)C1=CC=CC=C1

Specifications

Melting Point 114°C to 120°C
Quantity 25 g
Beilstein 639757
Formula Weight 178.19
Percent Purity ≥98%
Chemical Name or Material 3-Benzoylpropionic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 218-135-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only