missing translation for 'onlineSavingsMsg'
Learn More

3-Benzoyl-1,1,1-trifluoroacetone, 98+%

Catalog No. AAA1711022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1711022 100 g
AAA1711014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1711022 Supplier Thermo Scientific Chemicals Supplier No. A1711022

CAS: 326-06-7 | C10H7F3O2 | 216.16 g/mol

3-Benzoyl-1,1,1-trifluoroacetone, is used as a primary and secondary intermediate. It is used as an Intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Benzoyl-1,1,1-trifluoroacetone, is used as a primary and secondary intermediate. It is used as an Intermediate in organic synthesis.

Solubility
Sparingly Soluble in water (0.24 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.

Chemical Identifiers

CAS 326-06-7
Molecular Formula C10H7F3O2
Molecular Weight (g/mol) 216.16
MDL Number MFCD00000425
InChI Key VVXLFFIFNVKFBD-UHFFFAOYSA-N
Synonym benzoyl-1,1,1-trifluoroacetone, benzoyltrifluoroacetone, 4,4,4-trifluoro-1-phenyl-1,3-butanedione, benzoyl trifluoroacetyl methane, 3-benzoyl-1,1,1-trifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-phenyl, 4,4,4-trifluoro-1-phenyl-butane-1,3-dione, 1-benzoyl-3,3,3-trifluoroacetone, .omega.-trifluoroacetyl acetophenone, 1-benzoyl-3,3,3-trifluoro-2-propanone
PubChem CID 67589
IUPAC Name 4,4,4-trifluoro-1-phenylbutane-1,3-dione
SMILES FC(F)(F)C(=O)CC(=O)C1=CC=CC=C1

Specifications

Melting Point 37°C to 40°C
Density 1.113
Boiling Point 224°C
Flash Point 98°C (208°F)
Quantity 100 g
Beilstein 1875083
Solubility Information Sparingly Soluble in water (0.24g/L) (25°C).
Formula Weight 216.16
Percent Purity ≥98%
Chemical Name or Material 3-Benzoyl-1,1,1-trifluoroacetone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-307-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only