missing translation for 'onlineSavingsMsg'
Learn More

3-Benzalbutyramide 98.0+%, TCI America™

Catalog No. B08925G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B08925G Supplier TCI America Supplier No. B08925G
Only null left

Chemical Identifiers

CAS 7236-47-7
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.23
MDL Number MFCD00059860
InChI Key KAJZGRFYZKWYDX-VQHVLOKHSA-N
Synonym 3-Benzylidenebutyramide
PubChem CID 5371849
IUPAC Name (3E)-3-methyl-4-phenylbut-3-enamide
SMILES C\C(CC(N)=O)=C/C1=CC=CC=C1

Specifications

Melting Point 132°C
Color White
Quantity 5 g
Formula Weight 175.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Benzalbutyramide