missing translation for 'onlineSavingsMsg'
Learn More

3'-Azido-3'-deoxythymidine 98.0+%, TCI America™

Catalog No. p-7117700
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20521G 1 g
A20525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20521G Supplier TCI America Supplier No. A20521G

Chemical Identifiers

CAS 30516-87-1
Molecular Formula C10H13N5O4
Molecular Weight (g/mol) 267.25
MDL Number MFCD00006536
InChI Key HBOMLICNUCNMMY-CFQLRCIDNA-N
Synonym azt; azidothymidine; zdv; zidovudine, 1-2r-4-azido-5-hydroxymethyl oxolan-2-yl-5-methyl-3h-pyrimidine-2,4-dione
PubChem CID 455007
IUPAC Name 1-[(2R,4S,5S)-4-azido-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES CC1=CN([C@H]2C[C@H](N=[N+]=[N-])[C@@H](CO)O2)C(=O)NC1=O

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 1 g
Formula Weight 267.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Azido-3′-deoxythymidine
RTECSNumber XP2072000
TSCA No