missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 3'-Azido-3'-deoxythymidine, 98%

Catalog No. p-7043026
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6512703 1 g
AAJ6512706 5 g
AAJ6512714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6512703 Supplier Thermo Scientific Chemicals Supplier No. J6512703

Anti-viral agent | CAS: 30516-87-1 | C10H13N5O4 | 267.25 g/mol

3'-Azido-3'-deoxythymidine is a Anti-viral agent, and also used as a reverse transcriptase inhibitor active against HIV-1 virus that blocks the incorporation of nucleotides into growing DNA strands. Useful for antiviral and anticancer studies. 3?-Azido-3?-deoxythymidine enters CNS by diffusion and causes irreversible telomere shortening.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′-Azido-3′-deoxythymidine is a Anti-viral agent, and also used as a reverse transcriptase inhibitor active against HIV-1 virus that blocks the incorporation of nucleotides into growing DNA strands. Useful for antiviral and anticancer studies. 3′-Azido-3′-deoxythymidine enters CNS by diffusion and causes irreversible telomere shortening.

Solubility
Soluble in water.

Notes
Light Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 30516-87-1
Molecular Formula C10H13N5O4
Molecular Weight (g/mol) 267.25
MDL Number MFCD00006536
InChI Key HBOMLICNUCNMMY-CFQLRCIDNA-N
Synonym azt; azidothymidine; zdv; zidovudine, 1-2r-4-azido-5-hydroxymethyl oxolan-2-yl-5-methyl-3h-pyrimidine-2,4-dione
PubChem CID 455007
SMILES CC1=CN([C@H]2C[C@H](N=[N+]=[N-])[C@@H](CO)O2)C(=O)NC1=O

Specifications

Melting Point 113°C to 115°C
Color White
Quantity 1 g
Beilstein 3595791
Sensitivity Light sensitive; Very Hygroscopic
Merck Index 1410123
Solubility Information Soluble in water.
IUPAC Name 1-[(2R,5S)-4-azido-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
Formula Weight 267.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3'-Azido-3'-deoxythymidine
Show More Show Less
Hazard Category Serious eye damage/eye irritation (category 2), Germ cell mutagenicity (category 2), Specific target organ toxicity after single exposure (category 3), Carcinogenicity (category 2), Skin corrosion/irritation (category 2)
Hazard Statement Causes serious eye irritation, Suspected of causing genetic defects, May cause respiratory irritation, Suspected of causing cancer, Causes skin irritation
RTECSNumber XP207200
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only