Learn More
3-Azido-1-propylamine
Azido-containing reagent with carbonyl and carboxyl reactivity | CAS: 88192-19-2 | C3H8N4 | 100.125 g/mol
Supplier: Thermo Scientific Chemicals J6566803
Description
Azido-containing reagent with carbonyl and carboxyl reactivity3-Azido-1-propylamine is used in the preparation of perylenedimides derivative by reacting with 1,6,7,12-tetrachloroperylene-3,4:9,10-tetracarboxyanhydride, which is used in click chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAzido-containing reagent with carbonyl and carboxyl reactivity3-Azido-1-propylamine is used in the preparation of perylenedimides derivative by reacting with 1,6,7,12-tetrachloroperylene-3,4:9,10-tetracarboxyanhydride, which is used in click chemistry.
Solubility
Soluble in DMSO, DMF, DCM, chloroform, THF, and water.Miscible with dimethylsulfoxide, dimethyl formamide, dichloromethane, tetrahydrofuran, chloroform and water.
Notes
Incompatible with strong oxidizing agents.
Specifications
| 3-Azido-1-propylamine | |
| Yellow | |
| C3H8N4 | |
| MFCD11046568 | |
| UN1993 | |
| Soluble in DMSO,DMF,DCM,chloroform,THF,and water;Miscible with dimethylsulfoxide,dimethyl formamide,dichloromethane,tetrahydrofuran,chloroform and water. | |
| C(CN)CN=[N+]=[N-] | |
| 100.125 | |
| 100.12 |
| 88192-19-2 | |
| 60°C (140°F) | |
| N3CH2CH2CH2NH2 | |
| 1 g | |
| 3-azidopropylamine, 3-azido-1-propanamine, 1-propanamine, 3-azido, 3-azido-propylamine, 1-azido-3-aminopropane, 1-amino-3-azidopropane, 3-aminopropyl azide, 3-azido-1-propylamine, 3-azidopropyl-1-amine | |
| OYBOVXXFJYJYPC-UHFFFAOYSA-N | |
| 3-azidopropan-1-amine | |
| 150110 | |
| Liquid |
Chemical Identifiers
| 88192-19-2 | |
| 100.125 | |
| OYBOVXXFJYJYPC-UHFFFAOYSA-N | |
| 150110 | |
| C(CN)CN=[N+]=[N-] |
| C3H8N4 | |
| MFCD11046568 | |
| 3-azidopropylamine, 3-azido-1-propanamine, 1-propanamine, 3-azido, 3-azido-propylamine, 1-azido-3-aminopropane, 1-amino-3-azidopropane, 3-aminopropyl azide, 3-azido-1-propylamine, 3-azidopropyl-1-amine | |
| 3-azidopropan-1-amine |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only