missing translation for 'onlineSavingsMsg'
Learn More

3-Azetidinemethanol hydrochloride, 95%

Catalog No. AAH57801ME
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57801ME 500 mg
AAH57801MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH57801ME Supplier Thermo Scientific Chemicals Supplier No. H57801ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 928038-44-2 | C4H10ClNO | 123.58 g/mol

3-Azetidinemethanol Hydrochloride is used in the synthesis of human rhinovirus inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Azetidinemethanol Hydrochloride is used in the synthesis of human rhinovirus inhibitors.

Notes
Hygroscopic. Store at room temperature.

Chemical Identifiers

CAS 928038-44-2
Molecular Formula C4H10ClNO
Molecular Weight (g/mol) 123.58
MDL Number MFCD08703352
InChI Key AQUVQGSNKVDBBF-UHFFFAOYSA-N
Synonym azetidin-3-ylmethanol hydrochloride, 3-azetidinemethanol hydrochloride, azetidine-3-yl methanol hcl, 3-hydroxymethyl-azetidine hcl, azetidin-3-yl methanol hydrochloride, 3-hydroxymethyl azetidine hydrochloride, 3-azetidinemethanol, hydrochloride 1:1, azetidine-3-yl methanol hydrochloride, pubchem19094, 3-hydroxymethyl azetidine 3
PubChem CID 24801400
IUPAC Name azetidin-3-ylmethanol;hydrochloride
SMILES C1C(CN1)CO.Cl

Specifications

Quantity 500 mg
Sensitivity Hygroscopic
Formula Weight 123.58
Percent Purity 95%
Chemical Name or Material 3-Azetidinemethanol hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only