missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Anilinopropionitrile 98.0+%, TCI America™
$149.13 - $1260.10
Chemical Identifiers
| CAS | 1075-76-9 |
|---|---|
| Molecular Formula | C9H10N2 |
| Molecular Weight (g/mol) | 146.193 |
| MDL Number | MFCD00001953 |
| InChI Key | FENJKTQEFUPECW-UHFFFAOYSA-N |
| Synonym | 3-anilinopropionitrile, 3-phenylamino propanenitrile, n-2-cyanoethyl aniline, propanenitrile, 3-phenylamino, beta-anilinopropionitrile, 2-phenylaminopropionitrile, n-cyanoethyl aniline, propionitrile, 3-anilino, propanenitrile, 3-anilino |
| PubChem CID | 14100 |
| IUPAC Name | 3-anilinopropanenitrile |
| SMILES | C1=CC=C(C=C1)NCCC#N |
Chemical Identifiers
| 1075-76-9 | |
| 146.193 | |
| FENJKTQEFUPECW-UHFFFAOYSA-N | |
| 14100 | |
| C1=CC=C(C=C1)NCCC#N |
| C9H10N2 | |
| MFCD00001953 | |
| 3-anilinopropionitrile, 3-phenylamino propanenitrile, n-2-cyanoethyl aniline, propanenitrile, 3-phenylamino, beta-anilinopropionitrile, 2-phenylaminopropionitrile, n-cyanoethyl aniline, propionitrile, 3-anilino, propanenitrile, 3-anilino | |
| 3-anilinopropanenitrile |
Specifications
| 1075-76-9 | |
| White-Yellow | |
| C9H10N2 | |
| 25 g | |
| FENJKTQEFUPECW-UHFFFAOYSA-N | |
| 3-anilinopropanenitrile | |
| 14100 | |
| ≥98.0% (GC,T) | |
| 3-Anilinopropionitrile |
| 50°C | |
| 160°C | |
| MFCD00001953 | |
| 3-anilinopropionitrile, 3-phenylamino propanenitrile, n-2-cyanoethyl aniline, propanenitrile, 3-phenylamino, beta-anilinopropionitrile, 2-phenylaminopropionitrile, n-cyanoethyl aniline, propionitrile, 3-anilino, propanenitrile, 3-anilino | |
| C1=CC=C(C=C1)NCCC#N | |
| 146.193 | |
| 146.19 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-0122
RTECSNumber : UG0770000
TSCA : Yes