missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Anilinopropionitrile 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 50°C |
| Color | White-Yellow |
| Boiling Point | 160°C |
| Quantity | 500 g |
| Formula Weight | 146.19 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Anilinopropionitrile |
Safety and Handling
| EINECSNumber | (3)-0122 |
| RTECSNumber | UG0770000 |
| TSCA | Yes |