missing translation for 'onlineSavingsMsg'
Learn More

3-Anilinopropionitrile 98.0+%, TCI America™

Catalog No. A047725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A047725G 25 g
A0477500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A047725G Supplier TCI America Supplier No. A047725G

Chemical Identifiers

CAS 1075-76-9
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.193
MDL Number MFCD00001953
InChI Key FENJKTQEFUPECW-UHFFFAOYSA-N
Synonym 3-anilinopropionitrile, 3-phenylamino propanenitrile, n-2-cyanoethyl aniline, propanenitrile, 3-phenylamino, beta-anilinopropionitrile, 2-phenylaminopropionitrile, n-cyanoethyl aniline, propionitrile, 3-anilino, propanenitrile, 3-anilino
PubChem CID 14100
IUPAC Name 3-anilinopropanenitrile
SMILES C1=CC=C(C=C1)NCCC#N

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 160°C
Quantity 25 g
Formula Weight 146.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Anilinopropionitrile
EINECSNumber (3)-0122
RTECSNumber UG0770000
TSCA Yes