missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminotetrahydrofuran 97.0+%, TCI America™
Supplier: TCI America A27001G
Specifications
| 3-Aminotetrahydrofuran | |
| Colorless | |
| C4H9NO | |
| 1 g | |
| MIPHRQMEIYLZFZ-UHFFFAOYSA-N | |
| oxolan-3-amine | |
| 3365553 | |
| ≥97.0% (GC) |
| 88675-24-5 | |
| 73°C | |
| MFCD07778394 | |
| 1993 | |
| C1COCC1N | |
| 87.122 | |
| 87.12 | |
| Liquid |
Chemical Identifiers
| 88675-24-5 | |
| 87.122 | |
| MIPHRQMEIYLZFZ-UHFFFAOYSA-N | |
| oxolan-3-amine |
| C4H9NO | |
| MFCD07778394 | |
| 3365553 | |
| C1COCC1N |
Safety and Handling
TSCA : No