missing translation for 'onlineSavingsMsg'
Learn More

3-Aminoquinoline 99.0+%, TCI America™

Catalog No. A04185G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A04185G 5 g
A041825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A04185G Supplier TCI America Supplier No. A04185G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 580-17-6
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD00006772
InChI Key SVNCRRZKBNSMIV-UHFFFAOYSA-N
Synonym 3-aminoquinoline, 3-quinolinamine, 3-quinolylamine, 3-quinolineamine, quinoline, 3-amino, quinolin-3-ylamine, 3-amino quinoline, unii-98ovl5k3r4, quinolin-3-yl-amine, ccris 1678
PubChem CID 11375
IUPAC Name quinolin-3-amine
SMILES C1=CC=C2C(=C1)C=C(C=N2)N

Specifications

Melting Point 93°C
Boiling Point 137°C
Quantity 5 g
Formula Weight 144.18
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminoquinoline
RTECSNumber VA9622000
TSCA Yes