missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopyrazine-2-carboxamide, 96%

Catalog No. AAH6355303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6355303 1 g
AAH6355306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6355303 Supplier Thermo Scientific Chemicals Supplier No. H6355303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 32587-10-3 | C5H6N4O | 138.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32587-10-3
Molecular Formula C5H6N4O
Molecular Weight (g/mol) 138.13
MDL Number MFCD03407446
InChI Key SFSMATGDLFHTHE-UHFFFAOYSA-N
Synonym aminopyrazine carboxamide, 3-aminopyrazinecarboxamide, 3-amino-2-pyrazinecarboxamide, 3-amino-pyrazine-2-carboxylic acid amide, 3-amino-pyrazinecarboxamide, 3-amino-pyrazine-2-carboxamide, 2-pyrazinecarboxamide,3-amino, pyrazinecarboxamide, 3-amino, pyrazinecarboxamide, 3-amino-7ci,9ci
PubChem CID 280292
IUPAC Name 3-aminopyrazine-2-carboxamide
SMILES NC(=O)C1=NC=CN=C1N

Specifications

Quantity 1 g
Formula Weight 138.13
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Aminopyrazine-2-carboxamide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only