missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropyltrimethoxysilane 96.0+%, TCI America™

Catalog No. T1255500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1255500ML 500 mL
T125525ML 25 mL
T1255100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T1255500ML Supplier TCI America Supplier No. T1255500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13822-56-5
Molecular Formula C6H17NO3Si
Molecular Weight (g/mol) 179.291
MDL Number MFCD00008206
InChI Key SJECZPVISLOESU-UHFFFAOYSA-N
Synonym 3-aminopropyltrimethoxysilane, 3-aminopropyl trimethoxysilane, 3-trimethoxysilyl-1-propanamine, 3-trimethoxysilyl propan-1-amine, 3-trimethoxysilyl propylamine, 1-propanamine, 3-trimethoxysilyl, silane sc 3900, n-trimethoxysilylpropyl amine, propylamine, 3-trimethoxysilyl, unii-7mry0c9sb6
PubChem CID 83756
IUPAC Name 3-trimethoxysilylpropan-1-amine
SMILES CO[Si](CCCN)(OC)OC

Specifications

Color Yellow
Boiling Point 92°C
Quantity 500 mL
UN Number 1760
Formula Weight 179.29
Percent Purity ≥96.0% (GC,T)
Grade Reagent
Physical Form Liquid
Chemical Name or Material 3-Aminopropyltrimethoxysilane
EINECSNumber (2)-2061
TSCA Yes