missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropyltrimethoxysilane, 95%

CAS: 13822-56-5 | C6H17NO3Si | 179.29 g/mol

$131.52 - $131.52

Chemical Identifiers

CAS 13822-56-5
Molecular Formula C6H17NO3Si
Molecular Weight (g/mol) 179.29
MDL Number MFCD00008206
InChI Key SJECZPVISLOESU-UHFFFAOYSA-N
Synonym 3-aminopropyltrimethoxysilane, 3-aminopropyl trimethoxysilane, 3-trimethoxysilyl-1-propanamine, 3-trimethoxysilyl propan-1-amine, 3-trimethoxysilyl propylamine, 1-propanamine, 3-trimethoxysilyl, silane sc 3900, n-trimethoxysilylpropyl amine, propylamine, 3-trimethoxysilyl, unii-7mry0c9sb6
PubChem CID 83756
IUPAC Name 3-trimethoxysilylpropan-1-amine
SMILES CO[Si](CCCN)(OC)OC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC313251000
SDS View Documents
Thermo Scientific Chemicals
313251000
100 mL Glass bottle
Each for $131.52
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13822-56-5
179.29
SJECZPVISLOESU-UHFFFAOYSA-N
83756
CO[Si](CCCN)(OC)OC
C6H17NO3Si
MFCD00008206
3-aminopropyltrimethoxysilane, 3-aminopropyl trimethoxysilane, 3-trimethoxysilyl-1-propanamine, 3-trimethoxysilyl propan-1-amine, 3-trimethoxysilyl propylamine, 1-propanamine, 3-trimethoxysilyl, silane sc 3900, n-trimethoxysilylpropyl amine, propylamine, 3-trimethoxysilyl, unii-7mry0c9sb6
3-trimethoxysilylpropan-1-amine

Specifications

13822-56-5
100.0
1.0100g/mL
92°C
94% min. (GC)
C6H17NO3Si
NH2(CH2)3Si(OCH3)3
100 mL
3-aminopropyltrimethoxysilane, 3-aminopropyl trimethoxysilane, 3-trimethoxysilyl-1-propanamine, 3-trimethoxysilyl propan-1-amine, 3-trimethoxysilyl propylamine, 1-propanamine, 3-trimethoxysilyl, silane sc 3900, n-trimethoxysilylpropyl amine, propylamine, 3-trimethoxysilyl, unii-7mry0c9sb6
SJECZPVISLOESU-UHFFFAOYSA-N
3-trimethoxysilylpropan-1-amine
83756
95%
3-Aminopropyltrimethoxysilane
94.0
Colorless
194.0°C
Authentic
Glass bottle
1.4230 to 1.4250
MFCD00008206
1.01
Solubility in water: reacts with water.
CO[Si](CCCN)(OC)OC
179.29
179.29
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye damage.

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.

GHS Signal Word: Danger

EINECSNumber : 237-511-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only