missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropyltrimethoxysilane, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 313250025

Catalog No. AC313250025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-Aminopropyltrimethoxysilane
94.0
1.0100g/mL
92°C
94% min. (GC)
C6H17NO3Si
NH2(CH2)3Si(OCH3)3
2.5L
3-aminopropyltrimethoxysilane, 3-aminopropyl trimethoxysilane, 3-trimethoxysilyl-1-propanamine, 3-trimethoxysilyl propan-1-amine, 3-trimethoxysilyl propylamine, 1-propanamine, 3-trimethoxysilyl, silane sc 3900, n-trimethoxysilylpropyl amine, propylamine, 3-trimethoxysilyl, unii-7mry0c9sb6
SJECZPVISLOESU-UHFFFAOYSA-N
3-trimethoxysilylpropan-1-amine
83756
95%
13822-56-5
100.0
194.0°C
Authentic
Glass bottle
1.4230 to 1.4250
MFCD00008206
1.01
Solubility in water: reacts with water.
CO[Si](CCCN)(OC)OC
179.29
179.29
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye damage.

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.

GHS Signal Word: Danger

EINECSNumber : 237-511-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only