missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropyltriethoxysilane 98.0+%, TCI America™

Catalog No. p-7116902
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A043925G 25 g
A0439100G 100 g
A0439500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A043925G Supplier TCI America Supplier No. A043925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 919-30-2
Molecular Formula C9H23NO3Si
Molecular Weight (g/mol) 221.37
MDL Number MFCD00008207,MFCD01324904
InChI Key WYTZZXDRDKSJID-UHFFFAOYSA-N
Synonym 3-aminopropyltriethoxysilane, 3-aminopropyl triethoxysilane, aptes, 3-triethoxysilyl propan-1-amine, 1-propanamine, 3-triethoxysilyl, silicone a-1100, silane 1100, 3-triethoxysilyl propylamine, propylamine, 3-triethoxysilyl, triethoxy 3-aminopropyl silane
PubChem CID 13521
IUPAC Name (3-aminopropyl)triethoxysilane
SMILES CCO[Si](CCCN)(OCC)OCC

Specifications

Color Colorless
Boiling Point 217°C
Quantity 25 g
UN Number 2735
Formula Weight 221.37
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Aminopropyltriethoxysilane
EINECSNumber (2)-2061
RTECSNumber TX2100000
TSCA Yes