missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropyltriethoxysilane, 99%, AcroSeal™

Catalog No. p-4228340
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430941000 100 mL AcroSeal™ Glass Bottle
AC430948000 800 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC430941000 Supplier Thermo Scientific Chemicals Supplier No. 430941000
Only null left

CAS: 919-30-2 | C9H23NO3Si | 221.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 919-30-2
Molecular Formula C9H23NO3Si
Molecular Weight (g/mol) 221.37
MDL Number MFCD00008207,MFCD01324904
InChI Key WYTZZXDRDKSJID-UHFFFAOYSA-N
Synonym 3-aminopropyltriethoxysilane, 3-aminopropyl triethoxysilane, aptes, 3-triethoxysilyl propan-1-amine, 1-propanamine, 3-triethoxysilyl, silicone a-1100, silane 1100, 3-triethoxysilyl propylamine, propylamine, 3-triethoxysilyl, triethoxy 3-aminopropyl silane
PubChem CID 13521
IUPAC Name 3-triethoxysilylpropan-1-amine
SMILES CCO[Si](CCCN)(OCC)OCC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -70.0°C
Density 0.9420g/mL
Boiling Point 217.0°C
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4200 to 1.4220
Linear Formula H2N(CH2)3Si(OC2H5)3
Quantity 100 mL
Specific Gravity 0.942
Solubility Information Solubility in water: reacts. Other solubilities: soluble in chloroform
Formula Weight 221.37
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Aminopropyltriethoxysilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 213-048-4

RUO – Research Use Only