missing translation for 'onlineSavingsMsg'
Learn More

(3-Aminopropyl)triethoxysilane, 98%

Catalog No. AAA1066822
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1066822 100 g
AAA1066836 500 g
AAA106680E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1066822 Supplier Thermo Scientific Chemicals Supplier No. A1066822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Alfa Aesar (3-Aminopropyl)triethoxysilane, 98%, Quantity: 100g, Boiling Point: 217 deg.C, CAS: 919-30-2, Molecular Formula: C9H23NO3Si, Molecular Weight: 221.372, Percent Purity: 98%, pH: 11 | CAS: 919-30-2 | C9H23NO3Si | 221.37 g/mol

(3-Aminopropyl)triethoxysilane is useful in the formation of aminopropyl derivative of glass. It acts as an adsorbent for affinity chromatography. It is used to prepare positively charged slides suitable for use with various immunohistochemical and in situ hybridization procedures. In addition, it is used as adhesives and sealant chemicals, paint additives and coating additives. Further, it is used as a silylation reagent for coating glass and silica surfaces and to crosslink and immobilize proteins and other molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Aminopropyl)triethoxysilane is useful in the formation of aminopropyl derivative of glass. It acts as an adsorbent for affinity chromatography. It is used to prepare positively charged slides suitable for use with various immunohistochemical and in situ hybridization procedures. In addition, it is used as adhesives and sealant chemicals, paint additives and coating additives. Further, it is used as a silylation reagent for coating glass and silica surfaces and to crosslink and immobilize proteins and other molecules.

Solubility
Miscible with toluene, acetone, chloroform and ethanol.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, acids and moisture.

Chemical Identifiers

CAS 919-30-2
Molecular Formula C9H23NO3Si
Molecular Weight (g/mol) 221.37
MDL Number MFCD00008207,MFCD01324904
InChI Key WYTZZXDRDKSJID-UHFFFAOYSA-N
Synonym 3-aminopropyltriethoxysilane, 3-aminopropyl triethoxysilane, aptes, 3-triethoxysilyl propan-1-amine, 1-propanamine, 3-triethoxysilyl, silicone a-1100, silane 1100, 3-triethoxysilyl propylamine, propylamine, 3-triethoxysilyl, triethoxy 3-aminopropyl silane
PubChem CID 13521
IUPAC Name 3-triethoxysilylpropan-1-amine
SMILES CCO[Si](CCCN)(OCC)OCC

Specifications

Boiling Point 217°C
pH 11
Quantity 100 g
Linear Formula H2N(CH2)3Si(OCH2CH3)3
UN Number UN2735
Beilstein 1754988
Flash Point 104°C (219°F)
Solubility Information Miscible with toluene,acetone,chloroform and ethanol.
Formula Weight 221.37
Percent Purity 98%
Odor Amine-like
Refractive Index 1.421
Sensitivity Air and moisture sensitive
Density 0.948
Chemical Name or Material (3-Aminopropyl)triethoxysilane
Show More Show Less
Hazard Category H302-H314-H317-H335
Hazard Statement GHS H Statement
H314-H318-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 213-048-4
RTECSNumber TX2100000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only