missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich (3-Aminopropyl)triethoxysilane

Catalog No. 440140500ML
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
440140500ML 500 mL
440140100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 440140500ML Supplier Sigma Aldrich Supplier No. 440140500ML
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(3-Aminopropyl)tiethoxysilane (APTES) is used as an amino-silane which is mainly used as a dispersant. APTES attaches an amino group to the functional silane for bio-conjugation.Used in a preparation of a series of tenary composite films of polyimide/SiO/polydiphenylsiloxane with good optical transparency.

Specifications

CAS 919-30-2
Density 0.946 g/mL (at 25°C)
Boiling Point 217°C (760 mmHg)
Molecular Formula C9H23NO3Si
Linear Formula H2N(CH2)3 Si(OC2H5)3
MDL Number MFCD00008207
Quantity 500 mL
Synonym 3-Triethoxysilylpropylamine; APTES; APTS
Molecular Weight (g/mol) 221.37
Percent Purity 99%