Learn More
(3-Aminopropyl)diethoxymethylsilane, 97%
CAS: 3179-76-8 | C8H21NO2Si | 191.346 g/mol
$59.66 - $142.99
Chemical Identifiers
| CAS | 3179-76-8 |
|---|---|
| Molecular Formula | C8H21NO2Si |
| Molecular Weight (g/mol) | 191.346 |
| MDL Number | MFCD00039785 |
| InChI Key | HXLAEGYMDGUSBD-UHFFFAOYSA-N |
| Synonym | 3-aminopropylmethyldiethoxysilane, 3-diethoxymethylsilyl propylamine, dynasylan 1506, 3-aminopropyl-methyl-diethoxysilane, 1-propanamine, 3-diethoxymethylsilyl, dynasylan 1505, 3-aminopropyl diethoxymethylsilane, 3-diethoxy methyl silyl propan-1-amine, unii-a36to38mqn, gamma-aminopropylmethyldiethoxysilane |
| PubChem CID | 18511 |
| IUPAC Name | 3-[diethoxy(methyl)silyl]propan-1-amine |
| SMILES | CCO[Si](C)(CCCN)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1640114
|
Thermo Scientific Chemicals
L1640114 |
25 g |
Each for $59.66
|
|
|||||
|
AAL1640122
|
Thermo Scientific Chemicals
L1640122 |
100 g |
Each for $142.99
|
|
|||||
Description
(3-Aminopropyl)diethoxymethylsilane is used as silane coupling agent. The aromatic imine monolayers were prepared by reacting (3-aminopropyl)diethoxymethylsilane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(3-Aminopropyl)diethoxymethylsilane is used as silane coupling agent. The aromatic imine monolayers were prepared by reacting (3-aminopropyl)diethoxymethylsilane.
Solubility
Soluble and reacts with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
| 3179-76-8 | |
| 191.346 | |
| HXLAEGYMDGUSBD-UHFFFAOYSA-N | |
| 18511 | |
| CCO[Si](C)(CCCN)OCC |
| C8H21NO2Si | |
| MFCD00039785 | |
| 3-aminopropylmethyldiethoxysilane, 3-diethoxymethylsilyl propylamine, dynasylan 1506, 3-aminopropyl-methyl-diethoxysilane, 1-propanamine, 3-diethoxymethylsilyl, dynasylan 1505, 3-aminopropyl diethoxymethylsilane, 3-diethoxy methyl silyl propan-1-amine, unii-a36to38mqn, gamma-aminopropylmethyldiethoxysilane | |
| 3-[diethoxy(methyl)silyl]propan-1-amine |
Specifications
| 197°C to 199°C | |
| 3179-76-8 | |
| C8H21NO2Si | |
| UN2735 | |
| 85°C (185°F) | |
| Soluble and reacts with water. | |
| CCO[Si](C)(CCCN)OCC | |
| 191.346 | |
| 191.35 | |
| 1.427 | |
| 0.92 |
| <-20°C | |
| 25 g | |
| MFCD00039785 | |
| 1744264 | |
| 3-aminopropylmethyldiethoxysilane, 3-diethoxymethylsilyl propylamine, dynasylan 1506, 3-aminopropyl-methyl-diethoxysilane, 1-propanamine, 3-diethoxymethylsilyl, dynasylan 1505, 3-aminopropyl diethoxymethylsilane, 3-diethoxy methyl silyl propan-1-amine, unii-a36to38mqn, gamma-aminopropylmethyldiethoxysilane | |
| HXLAEGYMDGUSBD-UHFFFAOYSA-N | |
| 3-[diethoxy(methyl)silyl]propan-1-amine | |
| 18511 | |
| 97% | |
| Air and moisture sensitive | |
| (3-Aminopropyl)diethoxymethylsilane |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 221-660-8
RTECSNumber : UI0700000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only