missing translation for 'onlineSavingsMsg'
Learn More

(3-Aminopropyl)diethoxymethylsilane, 97%

Catalog No. AAL1640114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1640114 25 g
AAL1640122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1640114 Supplier Thermo Scientific Chemicals Supplier No. L1640114

CAS: 3179-76-8 | C8H21NO2Si | 191.346 g/mol

(3-Aminopropyl)diethoxymethylsilane is used as silane coupling agent. The aromatic imine monolayers were prepared by reacting (3-aminopropyl)diethoxymethylsilane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Aminopropyl)diethoxymethylsilane is used as silane coupling agent. The aromatic imine monolayers were prepared by reacting (3-aminopropyl)diethoxymethylsilane.

Solubility
Soluble and reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 3179-76-8
Molecular Formula C8H21NO2Si
Molecular Weight (g/mol) 191.346
MDL Number MFCD00039785
InChI Key HXLAEGYMDGUSBD-UHFFFAOYSA-N
Synonym 3-aminopropylmethyldiethoxysilane, 3-diethoxymethylsilyl propylamine, dynasylan 1506, 3-aminopropyl-methyl-diethoxysilane, 1-propanamine, 3-diethoxymethylsilyl, dynasylan 1505, 3-aminopropyl diethoxymethylsilane, 3-diethoxy methyl silyl propan-1-amine, unii-a36to38mqn, gamma-aminopropylmethyldiethoxysilane
PubChem CID 18511
IUPAC Name 3-[diethoxy(methyl)silyl]propan-1-amine
SMILES CCO[Si](C)(CCCN)OCC

Specifications

Boiling Point 197°C to 199°C
Melting Point <-20°C
Quantity 25 g
UN Number UN2735
Beilstein 1744264
Flash Point 85°C (185°F)
Solubility Information Soluble and reacts with water.
Formula Weight 191.35
Percent Purity 97%
Refractive Index 1.427
Sensitivity Air and moisture sensitive
Density 0.92
Chemical Name or Material (3-Aminopropyl)diethoxymethylsilane
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 221-660-8
RTECSNumber UI0700000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only