missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropane-1-sulfonic acid, 97%

Catalog No. p-7037231
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5591803 1 g
AAH5591806 5 g
AAH5591814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5591803 Supplier Thermo Scientific Chemicals Supplier No. H5591803
Only null left

CAS: 3687-18-1 | C3H9NO3S | 139.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3687-18-1
Molecular Formula C3H9NO3S
Molecular Weight (g/mol) 139.17
MDL Number MFCD00008225
InChI Key SNKZJIOFVMKAOJ-UHFFFAOYSA-N
Synonym homotaurine, 3-amino-1-propanesulfonic acid, tramiprosate, alzhemed, 3-aminopropanesulphonic acid, 3aps, 1-propanesulfonic acid, 3-amino, cerebril, 3-aminopropanesulfonic acid, 3-aminopropane sulfonic acid
PubChem CID 1646
ChEBI CHEBI:1457
IUPAC Name 3-aminopropane-1-sulfonic acid
SMILES NCCCS(O)(=O)=O

Specifications

Melting Point 293°C (decomposition)
Linear Formula H2N(CH2)3SO3H
Quantity 1 g
Merck Index 14,10218
Formula Weight 139.17
Percent Purity 97%
Chemical Name or Material 3-Aminopropane-1-sulfonic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-977-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only