missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopiperidine dihydrochloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 398110010

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC398110010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

138060-07-8
173.09
GGPNYXIOFZLNKW-UHFFFAOYSA-N
16211870
C1CC(CNC1)N.Cl.Cl
C5H12N2·2HCl
MFCD00012773
3-aminopiperidine dihydrochloride, piperidin-3-amine dihydrochloride, 3-aminopiperidine 2hcl, 3-aminopiperidine hcl, 3-piperidinamine dihydrochloride, 3-piperidinamine, dihydrochloride, pubchem6755, acmc-209tvl, ksc839o8j, 3-amino-piperidine di-hydrochloride
piperidin-3-amine;dihydrochloride

Specifications

3-Aminopiperidine dihydrochloride
96.0
Authentic
Glass bottle
1g
3-aminopiperidine dihydrochloride, piperidin-3-amine dihydrochloride, 3-aminopiperidine 2hcl, 3-aminopiperidine hcl, 3-piperidinamine dihydrochloride, 3-piperidinamine, dihydrochloride, pubchem6755, acmc-209tvl, ksc839o8j, 3-amino-piperidine di-hydrochloride
C1CC(CNC1)N.Cl.Cl
173.09
173.09
138060-07-8
100.0
97%
C5H12N2·2HCl
MFCD00012773
GGPNYXIOFZLNKW-UHFFFAOYSA-N
piperidin-3-amine;dihydrochloride
16211870
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents