missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophthalhydrazide monosodium salt, 98+%

CAS: 20666-12-0 | C8H9N3NaO2 | 202.169 g/mol

Supplier:  Thermo Scientific Chemicals L1520506

 View more versions of this product

Catalog No. AAL1520506


Only null left
Explore available promotions

Description

Description

3-Aminophthalhydrazide monosodium salt is used for chemiluminescence analysis of metal cations, blood and glucococorticoids. It is also to image myeloperoxidase activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminophthalhydrazide monosodium salt is used for chemiluminescence analysis of metal cations, blood and glucococorticoids. It is also to image myeloperoxidase activity.

Solubility
Soluble in water (50 mg/ml).

Notes
Incompatible with bases, reducing agents and oxidizing agents. It is hygroscopic in nature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

3-Aminophthalhydrazide monosodium salt
C8H9N3NaO2
5 g
luminol sodium salt
BTPIPBDXONOBET-UHFFFAOYSA-N
5-amino-2,3-dihydrophthalazine-1,4-dione;molecular hydrogen;sodium
131673872
≥98%
20666-12-0
MFCD00161491
Hygroscopic
Soluble in water (50mg/ml).
[HH].C1=CC2=C(C(=C1)N)C(=O)NNC2=O.[Na]
202.169
199.15

Chemical Identifiers

20666-12-0
202.169
BTPIPBDXONOBET-UHFFFAOYSA-N
131673872
[HH].C1=CC2=C(C(=C1)N)C(=O)NNC2=O.[Na]
C8H9N3NaO2
MFCD00161491
luminol sodium salt
5-amino-2,3-dihydrophthalazine-1,4-dione;molecular hydrogen;sodium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 629-344-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only