missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophthalhydrazide monosodium salt, 98+%

Catalog No. AAL1520503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1520503 1 g
AAL1520506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1520503 Supplier Thermo Scientific Chemicals Supplier No. L1520503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20666-12-0 | C8H9N3NaO2 | 202.169 g/mol

3-Aminophthalhydrazide monosodium salt is used for chemiluminescence analysis of metal cations, blood and glucococorticoids. It is also to image myeloperoxidase activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminophthalhydrazide monosodium salt is used for chemiluminescence analysis of metal cations, blood and glucococorticoids. It is also to image myeloperoxidase activity.

Solubility
Soluble in water (50 mg/ml).

Notes
Incompatible with bases, reducing agents and oxidizing agents. It is hygroscopic in nature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 20666-12-0
Molecular Formula C8H9N3NaO2
Molecular Weight (g/mol) 202.169
MDL Number MFCD00161491
InChI Key BTPIPBDXONOBET-UHFFFAOYSA-N
Synonym luminol sodium salt
PubChem CID 131673872
IUPAC Name 5-amino-2,3-dihydrophthalazine-1,4-dione;molecular hydrogen;sodium
SMILES [HH].C1=CC2=C(C(=C1)N)C(=O)NNC2=O.[Na]

Specifications

Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Soluble in water (50mg/ml).
Formula Weight 199.15
Percent Purity ≥98%
Chemical Name or Material 3-Aminophthalhydrazide monosodium salt
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 629-344-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only