missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophthalhydrazide, 98%

Catalog No. AAA1459722
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1459722 100 g
AAA1459706 5 g
AAA1459714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1459722 Supplier Thermo Scientific Chemicals Supplier No. A1459722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Alfa Aesar 3-Aminophthalhydrazide, 98%, Quantity: 25g, Melting Point: 325 deg.C to 330 deg.C, Molecular Weight: 177.163, Percent Purity: 98%, Beilstein: 383929, CAS: 521-31-3, EINECS Number: 208-309-4, Formula Weight: 177.16, Health Hazard 1: H315-H319-H335 | CAS: 521-31-3 | C8H7N3O2 | 177.163 g/mol

3-Aminophthalhydrazide is used in the detection of copper, iron, peroxides and cyanides. It exhibits chemiluminescence and utilized to measure opsonic and phagocytic function. It acts as a biological sensor and involved in the detection of polymorphonuclear leukocytes response in a patient with a myeloperoxidase deficiency. Further, it is used as a forensic test for blood.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminophthalhydrazide is used in the detection of copper, iron, peroxides and cyanides. It exhibits chemiluminescence and utilized to measure opsonic and phagocytic function. It acts as a biological sensor and involved in the detection of polymorphonuclear leukocytes response in a patient with a myeloperoxidase deficiency. Further, it is used as a forensic test for blood.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.

Chemical Identifiers

CAS 521-31-3
Molecular Formula C8H7N3O2
Molecular Weight (g/mol) 177.163
MDL Number MFCD00006890
InChI Key HWYHZTIRURJOHG-UHFFFAOYSA-N
Synonym luminol, 3-aminophthalhydrazide, 3-aminophthalic hydrazide, 5-amino-2,3-dihydro-1,4-phthalazinedione, 1,4-phthalazinedione, 5-amino-2,3-dihydro, 3-aminophthalic acid hydrazide, 5-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione, unii-5exp385q4f, ccris 5962, 3-aminophthalylhydrazide
PubChem CID 10638
IUPAC Name 5-amino-2,3-dihydrophthalazine-1,4-dione
SMILES C1=CC2=C(C(=C1)N)C(=O)NNC2=O

Specifications

Melting Point 325°C to 330°C
Odor Odorless
Quantity 100 g
Beilstein 383929
Merck Index 14,5600
Solubility Information Soluble in water.
Formula Weight 177.16
Percent Purity 98%
Chemical Name or Material 3-Aminophthalhydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-309-4
RTECSNumber TH8890060
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only