missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophthalhydrazide, 98%, pure

Catalog No. AC153850010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC153850010 1 g Glass bottle
AC153850050 5 g Glass bottle
AC153850250 25 g Glass bottle
AC153851000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC153850010 Supplier Thermo Scientific Chemicals Supplier No. 153850010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

3-Aminophthalhydrazide, 98%, pure, Quantity: 25g, Packaging: Glass bottle, Blue-Purple to Green, Grade: Pure, Melting Point: 319.0 deg.C to 320.0 deg.C, Molecular Weight: 177.16, Percent Purity: 98%, Assay Percent Range: 98%, Beilstein: 25,IV,4192, CAS: 521-31-3, CAS Max %: 100.0 | CAS: 521-31-3 | C8H7N3O2 | 177.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 521-31-3
Molecular Formula C8H7N3O2
Molecular Weight (g/mol) 177.16
MDL Number MFCD00006890
InChI Key HWYHZTIRURJOHG-UHFFFAOYSA-N
Synonym luminol, 3-aminophthalhydrazide, 3-aminophthalic hydrazide, 5-amino-2,3-dihydro-1,4-phthalazinedione, 1,4-phthalazinedione, 5-amino-2,3-dihydro, 3-aminophthalic acid hydrazide, 5-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione, unii-5exp385q4f, ccris 5962, 3-aminophthalylhydrazide
PubChem CID 10638
IUPAC Name 5-amino-2,3-dihydrophthalazine-1,4-dione
SMILES C1=CC2=C(C(=C1)N)C(=O)NNC2=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 319.0°C to 320.0°C
Color Blue-Purple to Green
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Beilstein 25,IV,4192
Merck Index 15,566
Solubility Information (2% in 1 N NaOH) Clear yellow to yellow-green
Formula Weight 177.16
Percent Purity 98%
Grade Pure
Physical Form Powder
Chemical Name or Material 3-Aminophthalhydrazide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue
EINECSNumber 208-309-4
RTECSNumber TH8890060
TSCA TSCA

RUO – Research Use Only