missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminophenylboronic acid hydrochloride, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Color | White to Gray |
| Elemental Analysis | 40.52 to 42.60% C, 7.88 to 8.28% N |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% min. (HPLC) |
| Packaging | Glass Bottle |
| Quantity | 1 g |
| Formula Weight | 173.41 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Wash face,hands and any exposed skin thoroughly after handling. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre |
| EINECSNumber | 285-052-4 |
RUO – Research Use Only