missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophenylboronic Acid Hemisulfate, TCI America™

Catalog No. A17741G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A17741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A17741G Supplier TCI America Supplier No. A17741G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66472-86-4
Molecular Formula C12H18B2N2O8S
Molecular Weight (g/mol) 371.96
MDL Number MFCD00013111
InChI Key UKTAURVTSWDIQR-UHFFFAOYSA-N
Synonym 3-aminophenyl boronic acid sulfate 2:1, 3-aminobenzeneboronic acid hemisulfate salt, 3-aminophenylboronic acid hemisulfate, 3-aminobenzeneboronic acid hemisulfate, 3-aminophenylboronic acid hemisulphate, 4-aminophenylboronic acid hemisulfate, m-aminophenyl boronic acid, hemisulphate, boronic acid, 3-aminophenyl-, sulfate 2:1, 3-aminophenyl boronic acid; sulfuric acid, bis m-aminophenylboronic acid ; sulfuric acid
PubChem CID 16211139
IUPAC Name bis((3-aminophenyl)boronic acid); sulfuric acid
SMILES OS(O)(=O)=O.NC1=CC=CC(=C1)B(O)O.NC1=CC=CC(=C1)B(O)O

Specifications

Quantity 1 g
Formula Weight 185.98
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminophenylboronic Acid Hemisulfate