missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophenylboronic acid hemisulfate, 98%

Catalog No. AC104300250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104300250 25 g Glass bottle
AC104300050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC104300250 Supplier Thermo Scientific Chemicals Supplier No. 104300250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 66472-86-4 | C12H18B2N2O8S | 371.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 66472-86-4
Molecular Formula C12H18B2N2O8S
Molecular Weight (g/mol) 371.96
MDL Number MFCD00013111
InChI Key UKTAURVTSWDIQR-UHFFFAOYSA-N
Synonym 3-aminophenyl boronic acid sulfate 2:1, 3-aminobenzeneboronic acid hemisulfate salt, 3-aminophenylboronic acid hemisulfate, 3-aminobenzeneboronic acid hemisulfate, 3-aminophenylboronic acid hemisulphate, 4-aminophenylboronic acid hemisulfate, m-aminophenyl boronic acid, hemisulphate, boronic acid, 3-aminophenyl-, sulfate 2:1, 3-aminophenyl boronic acid; sulfuric acid, bis m-aminophenylboronic acid ; sulfuric acid
PubChem CID 16211139
SMILES OS(O)(=O)=O.NC1=CC=CC(=C1)B(O)O.NC1=CC=CC(=C1)B(O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2NC6H4B(OH)2·1/2H2SO4
Quantity 25 g
IUPAC Name (3-aminophenyl)boronic acid;sulfuric acid
Formula Weight 185.99
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminophenylboronic acid hemisulfate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 266-376-5

RUO – Research Use Only