missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopentane 98.0+%, TCI America™

Catalog No. A111525ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A111525ML 25 mL
A1115100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A111525ML Supplier TCI America Supplier No. A111525ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 616-24-0
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.17
MDL Number MFCD00008096
InChI Key PQPFFKCJENSZKL-UHFFFAOYSA-N
Synonym 3-aminopentane, 3-pentanamine, 1-ethylpropylamine, 3-pentylamine, propylamine, 1-ethyl, 3-amylamine, unii-3n2it605hv, 1-ethyl-propylamine, pent-3-ylamine, 3-amino pentane
PubChem CID 12019
ChEBI CHEBI:84248
IUPAC Name pentan-3-amine
SMILES CCC(N)CC

Specifications

Color Yellow
Boiling Point 91°C
Quantity 25 mL
UN Number 2733
Formula Weight 87.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Aminopentane
EINECSNumber (2)-0133
TSCA Yes